Learn More
CD23 Monoclonal Antibody (B-G6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen AHS2301
156.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.02000kg
Description
CD23 is a 45kDa glycoprotein, which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma.
CD23 Monoclonal antibody specifically detects CD23 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
CD23 | |
Monoclonal | |
Unconjugated | |
FCER2 | |
BLAST-2; CD23; CD23 antigen; CD23A; CLEC4J; C-type lectin domain family 4 member J; C-type lectin domain family 4, member J; Fc epsilon receptor II; FC epsilon RII; Fc fragment of IgE receptor II; Fc fragment of IgE, low affinity II, receptor for (CD23); Fc receptor alpha multiple ligand receptor; Fc receptor, IgE, low affinity II, alpha polypeptide; Fcalpha muR; Fcalpha RII; Fce2; fc-epsilon-RII; FCER2; Fcer2a; FceRII; IGEBF; Immunoglobulin E-binding factor; immunoglobulin epsilon-chain; low affinity immunoglobulin epsilon Fc receptor; Low affinity immunoglobulin epsilon Fc receptor membrane-bound form; Low affinity immunoglobulin epsilon Fc receptor soluble form; low-affinity IgE receptor; Ly-42; lymphocyte IgE receptor | |
Mouse | |
purified | |
RUO | |
2208 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry | |
B-G6 | |
PBS with 1% BSA and 0.09% sodium azide; pH 7.2 | |
P06734 | |
FCER2 | |
DAUDI (Burkitt cell line) | |
200 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |