missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD229 Monoclonal Antibody (HLy 9.1.25), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA135946
Code : LD
Additional Details : Weight : 0.01000kg
Description
The reagent is designed for analysis of human blood cells using 10 μL reagent/ 100 μL of whole blood or 10^6 cells in a suspension.
CD229 Monoclonal antibody specifically detects CD229 in Human samples. It is validated for Flow Cytometry.Specifications
CD229 | |
Monoclonal | |
PE | |
Ly9 | |
AI893573; CD229; CDABP0070; Cell surface molecule Ly-9; hly9; Lgp100; LY9; Ly-9; Lymphocyte antigen 9; mLY9; RP11-312J18.1; Signaling lymphocytic activation molecule 3; SLAM family member 3; SLAMF3; T100; T-lymphocyte surface antigen Ly-9 | |
Mouse | |
purified | |
RUO | |
4063 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
HLy 9.1.25 | |
PBS with 1% BSA and 0.09% sodium azide | |
Q9HBG7 | |
Ly9 | |
Mouse pre-B cell line 300.19 transfected with CD229 cDNA. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |