missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD223 Monoclonal Antibody (631501), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524284
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
CD223 Monoclonal antibody specifically detects CD223 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD223 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q61790 | |
LAG3 | |
NS0 mouse myeloma cell line transfected with mouse LAG-3 Ser23-Leu442 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry | |
631501 | |
Unconjugated | |
LAG3 | |
Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3 | |
Rat | |
Protein A/G | |
RUO | |
16768 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |