missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527928
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD22 Polyclonal antibody specifically detects CD22 in Human samples. It is validated for Western Blot.Specifications
CD22 | |
Polyclonal | |
Unconjugated | |
CD22 | |
A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
933 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P20273 | |
CD22 | |
Recombinant fragment corresponding to a region within amino acids 193 and 710 of Human CD22 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |