Learn More
CD22 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547292
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) Siglec-2 is observed and less than 1% cross-reactivity with rhSiglec-5, rhSiglec-7, rhSiglec-9, rhSiglec-10, recombinant mouse (rm) Siglec-3, and rmSiglec-F is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CD22 Polyclonal antibody specifically detects CD22 in Mouse samples. It is validated for Immunocytochemistry, Neutralization, Western Blot.Specifications
CD22 | |
Polyclonal | |
Unconjugated | |
CD22 | |
A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
12483 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P35329 | |
CD22 | |
Mouse myeloma cell line NS0-derived recombinant mouse Siglec-2/CD22 Ser22-Arg702 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |