missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Monoclonal Antibody (LT21), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119754
252.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LE
Additional Details : Weight : 0.01000kg
Description
CD21 Monoclonal antibody specifically detects CD21 in Bovine, Canine, Human, Porcine samples. It is validated for Flow Cytometry.Specifications
CD21 | |
Monoclonal | |
PE | |
CR2 | |
C3b/C4b receptor; C3-binding protein; C3BR; C3DR; C4BR; CD21; CD35; CD35 antigen; cell surface receptor for C3d; complement C3b/C4b receptor 1 (Knops blood group); complement C3d receptor; Complement C3d receptor (C3DR); complement C3d receptor 2; complement component (3b/4b) receptor 1 (Knops blood group); complement component (3d/Epstein Barr virus) receptor 2; complement component 3b/4b receptor 1 (Knops blood group); complement component 3d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; complement receptor type 1-like protein; complement receptor type 2; Complement Receptor type 2 (CR2); CR; CR1; Cr-1; CR1-L; Cr2; Cr-2; CVID7; EBV receptor; EBV-R; Epstein-Barr virus receptor; EVBR; expressed in B lymphocytes; KN; Knops blood group antigen; LOW QUALITY PROTEIN: complement receptor type 1; SLEB9 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
1380, 407126, 490269 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
LT21 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P20023 | |
CR2 | |
IM9 human B-lymphoblastoid cell line. | |
100 Tests | |
Primary | |
Bovine, Canine, Human, Porcine | |
Antibody | |
IgG1 |