Learn More
CD21 Monoclonal Antibody (CA2.1D6), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516609
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-16609 should not be used with MA5-16605 (mouse anti-canine CD3), in dual color flow cytometry, due to non-specific interactions between the two reagents. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. Mouse anti Canine CD21 antibody, clone CA2.1D6 recognizes canine CD21, also known as Complement receptor type 2. CD21 is a cell surface antigen expressed by canine B lymphocytes.
CD21 Monoclonal antibody specifically detects CD21 in Canine, Equine, Feline samples. It is validated for Flow Cytometry.Specifications
CD21 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
0 | |
CR2 | |
The details of the immunogen for this antibody are not currently available. | |
100 Tests | |
Primary | |
Canine, Equine, Feline | |
Antibody | |
IgG1 |
Flow Cytometry | |
CA2.1D6 | |
PE | |
CR2 | |
C3b/C4b receptor; C3-binding protein; C3BR; C3DR; C4BR; CD21; CD35; CD35 antigen; cell surface receptor for C3d; complement C3b/C4b receptor 1 (Knops blood group); complement C3d receptor; Complement C3d receptor (C3DR); complement C3d receptor 2; complement component (3b/4b) receptor 1 (Knops blood group); complement component (3d/Epstein Barr virus) receptor 2; complement component 3b/4b receptor 1 (Knops blood group); complement component 3d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; complement receptor type 1-like protein; complement receptor type 2; Complement Receptor type 2 (CR2); CR; CR1; Cr-1; CR1-L; Cr2; Cr-2; CVID7; EBV receptor; EBV-R; Epstein-Barr virus receptor; EVBR; expressed in B lymphocytes; KN; Knops blood group antigen; LOW QUALITY PROTEIN: complement receptor type 1; SLEB9 | |
Mouse | |
Protein G | |
RUO | |
101094393, 102149469, 490269 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |