Learn More
DC-SIGN Monoclonal Antibody (MR-1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516866
414.68 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This product is a low-endotoxin, preservative-free formulation. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
DC-SIGN Monoclonal antibody specifically detects DC-SIGN in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry.Specifications
DC-SIGN | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9NNX6 | |
CD209 | |
Immature myeloid monocyte-derived dendritic cells (MDDCs) | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay, Immunocytochemistry | |
MR-1 | |
Unconjugated | |
CD209 | |
CD209; CD209 antigen; CD209 antigen-like protein A; CD209 molecule; Cd209a; CD209a antigen; CD209a molecule; Cd209d; CD209d antigen; CDSIGN; Cire; CLEC4L; Clec4m; C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; C-type lectin domain family 4, member M; Dcsign; DC-SIGN; DC-SIGN1; Dc-signr; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin; dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein; MGC129965; MGC130443; RGD1561104; SIGN-R1; SIGNR5 | |
Mouse | |
Protein A | |
RUO | |
30835 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |