Learn More
CD206 Monoclonal Antibody (MR5D3), Biotin, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA516869
309.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CD206 is expressed weakly at the cell surface. Staining may be increased following membrane permeabilization. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti mouse CD206 antibody, clone MR5D3 recognizes the mouse mannose receptor, a approximately 175 kDa type 1 membrane glycoprotein that is also known as CD206.
CD206 Monoclonal antibody specifically detects CD206 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD206 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
Q61830 | |
Mrc1 | |
Chimaeric CRD4-7-Fc protein. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
MR5D3 | |
Biotin | |
Mrc1 | |
AW259686; bA541I19.1; CD206; CELE_Y39A3CR.4; CLEC13D; CLEC13DL; C-type lectin domain family 13 member D; C-type lectin domain family 13 member D-like; ddp-1; hMR; Human mannose receptor; macrophage mannose receptor 1; Macrophage mannose receptor 1-like protein 1; mannose receptor, C type 1; mannose receptor, C type 1-like 1; Mitochondrial import inner membrane translocase subunit Tim8; MMR; MR; MRC1; MRC1L1; tim-8; Y39A3CR.4 | |
Rat | |
Protein G | |
RUO | |
17533 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |