Learn More
CD205 Monoclonal Antibody (NLDC-145), FITC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA516890
Code : NEW
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse CD205, clone NLDC-145 recognizes DEC-205, a approximately 205 kDa integral membrane glycoprotein expressed by mouse dendritic cells and thymic epithelial cells.
CD205 (LY25, Lymphocyte Antigen 75) is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 acts as an endocytic receptor that directs captured antigens to the extracellular space towards a specialized antigen processing region. Further, CD205 also prompts the reduced proliferation of B lymphocytes. Diseases associated with CD205 dysfunction include pneumonic plague and lipid pneumonia.Specifications
CD205 | |
Monoclonal | |
0.1 mg/mL | |
Ly75 | |
RP23-10L17.4, CD205, DEC-205, ly-75, CLEC13B, GP200-MR6, DCL-1 fusion, LY75-CD302 fusion protein | |
Rat | |
Protein G | |
RUO | |
17076 | |
4°C or -20°C if preferred, store in dark | |
Liquid |
Flow Cytometry | |
NLDC-145 | |
FITC | |
Q60767 | |
LY75 | |
mouse lymphnode stroma emulsified in Freund′s complete adjuvant | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |