missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD203c Monoclonal Antibody (NP4D6), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110139
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD203c Monoclonal antibody specifically detects CD203c in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
CD203c | |
Monoclonal | |
PE | |
ENPP3 | |
AI876438; alkaline phosphodiesterase; Alkaline phosphodiesterase I; B10; CD203c; dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); ectonucleotide pyrophosphatase/phosphodiesterase 3; ectonucleotide pyrophosphatase/phosphodiesterase family member 3; E-NPP 3; ENPP3; gp130RB13-6; Npp3; NPPase; Nucleotide diphosphatase; Nucleotide pyrophosphatase; PD-Ibeta; Pdnp3; Phosphodiesterase I beta; phosphodiesterase I/nucleotide pyrophosphatase 3; phosphodiesterase-I beta; RB13-6; RB13-6 antigen | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
5169 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
NP4D6 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
O14638 | |
ENPP3 | |
HEK-293 cells transfected with human CD203c | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |