missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200 Monoclonal Antibody (OX-2), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517563
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CD200 Monoclonal antibody specifically detects CD200 in Rat samples. It is validated for Flow Cytometry.Specifications
CD200 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P04218 | |
Cd200 | |
Rat thymocyte membrane glycoproteins were fused with mouse NS-1 myeloma cells. | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
OX-2 | |
Biotin | |
Cd200 | |
antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein | |
Mouse | |
Protein G | |
RUO | |
24560 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |