missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200 Monoclonal Antibody (MRC OX104), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA135930
413.35 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CD200 Monoclonal antibody specifically detects CD200 in Human samples. It is validated for Flow Cytometry.Specifications
CD200 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
P41217 | |
Cd200 | |
Proprietary immunogen. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
MRC OX104 | |
FITC | |
Cd200 | |
antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein | |
Mouse | |
purified | |
RUO | |
4345 | |
4° C, store in dark | |
Liquid |