missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200 Monoclonal Antibody (MRC OX90), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA190713
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-90713 detects CD200 from mouse samples.
CD200 Monoclonal antibody specifically detects CD200 in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Frozen).Specifications
CD200 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O54901 | |
Cd200 | |
Mouse CD200-rat CD4 fusion protein | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen) | |
MRC OX90 | |
Unconjugated | |
Cd200 | |
antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein | |
Rat | |
Protein G | |
RUO | |
17470 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |