Learn More
CD20 Monoclonal Antibody (2H7), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA182403
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD20 antibody, clone 2H7 recognizes the human CD20 cell surface antigen, a 33-37 kDa non-glycosylated phosphoprotein.
CD20 Monoclonal antibody specifically detects CD20 in Human, Rhesus Monkey samples. It is validated for Flow Cytometry.Specifications
CD20 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
P11836 | |
Ms4a1 | |
Proprietary Immunogen. | |
100 Tests | |
Primary | |
Human, Rhesus Monkey | |
Antibody | |
IgG2b |
Flow Cytometry | |
2H7 | |
PE | |
Ms4a1 | |
AA960661; APY; ATOPY; B1; B-cell differentiation antigen Ly-44; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; Cd20; CD20 antigen; CD20 cell surface protein; CD20 receptor; CVID5; EGK_06167; Fc epsilon receptor I beta chain; Fc Fragment of IgE high affinity I receptor for beta polypeptide; FCER1B; High affinity immunoglobulin epsilon receptor subunit beta; IgE Fc receptor subunit beta; IGEL; IGER; IGHER; LEU16; LEU-16; Leukocyte surface antigen Leu-16; Ly44; Ly-44; Lymphocyte antigen 44; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; membrane-spanning 4-domains, subfamily A, member 2; MGC3969; MS4A1; MS4A2; S7 | |
Mouse | |
Protein A | |
RUO | |
696843, 931 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |