Learn More
CD20 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516701
Code : A0
Additional Details : Weight : 0.01000kg
Description
Upon delivery aliquot and store at -20oC, avoiding freeze/thaw cycles. For IHC staining, antigen retrieval is not essential but may optimize staining. Recommended positive controls: BJAB WCL and tonsil.
CD20 Polyclonal antibody specifically detects CD20 in Canine, Feline, Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD20 | |
Polyclonal | |
Unconjugated | |
Ms4a1 | |
AA960661; APY; ATOPY; B1; B-cell differentiation antigen Ly-44; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; Cd20; CD20 antigen; CD20 cell surface protein; CD20 receptor; CVID5; EGK_06167; Fc epsilon receptor I beta chain; Fc Fragment of IgE high affinity I receptor for beta polypeptide; FCER1B; High affinity immunoglobulin epsilon receptor subunit beta; IgE Fc receptor subunit beta; IGEL; IGER; IGHER; LEU16; LEU-16; Leukocyte surface antigen Leu-16; Ly44; Ly-44; Lymphocyte antigen 44; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; membrane-spanning 4-domains, subfamily A, member 2; MGC3969; MS4A1; MS4A2; S7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12482, 485430, 494216, 931 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.077 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P11836, P19437, Q3C2E2 | |
Ms4a1 | |
Synthetic peptide derived from C-terminus of human CD20 protein | |
500 μL | |
Primary | |
Canine, Feline, Human, Mouse | |
Antibody | |
IgG |