missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Monoclonal Antibody (2H7), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110169
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD20 Monoclonal antibody specifically detects CD20 in Human, Non-human Primate samples. It is validated for Flow Cytometry.Specifications
CD20 | |
Monoclonal | |
FITC | |
Ms4a1 | |
AA960661; APY; ATOPY; B1; B-cell differentiation antigen Ly-44; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; Cd20; CD20 antigen; CD20 cell surface protein; CD20 receptor; CVID5; EGK_06167; Fc epsilon receptor I beta chain; Fc Fragment of IgE high affinity I receptor for beta polypeptide; FCER1B; High affinity immunoglobulin epsilon receptor subunit beta; IgE Fc receptor subunit beta; IGEL; IGER; IGHER; LEU16; LEU-16; Leukocyte surface antigen Leu-16; Ly44; Ly-44; Lymphocyte antigen 44; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; membrane-spanning 4-domains, subfamily A, member 2; MGC3969; MS4A1; MS4A2; S7 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
931 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
2H7 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P11836 | |
Ms4a1 | |
Human tonsillar B cells | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG2b κ |