missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Monoclonal Antibody (LT19), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119595
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Specifications
CD19 | |
Monoclonal | |
FITC | |
Cd19 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
930 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
LT19 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P15391 | |
Cd19 | |
Daudi human Burkitt Lymphoma cell line | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |