missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Monoclonal Antibody (MB19-1), APC-Cyanine7, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15391
Code : NEW
Additional Details : Weight : 0.00280kg
Description
CD19 Monoclonal antibody specifically detects CD19 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD19 | |
Monoclonal | |
0.1 mg/mL | |
PBS with proprietary stabilizer and <0.1% sodium azide | |
P25918 | |
Cd19 | |
CD19+ mouse pre-B cell line, 300.191 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgA κ |
Flow Cytometry | |
MB19-1 | |
APC-Cyanine7 | |
Cd19 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Mouse | |
purified | |
RUO | |
12478 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |