Learn More
CD18 Monoclonal Antibody (WT.3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1817
Code : PN
Additional Details : Weight : 0.80000kg
Description
MA1817 targets CD18 in FACS, IP, and WB applications and shows reactivity with Rat samples. The MA1817 immunogen is rat beta2 Integrin. MA1817 detects CD18 which has a predicted molecular weight of approximately 83 kDa. This product is a Low Endotoxin formulation. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
CD18 Monoclonal antibody specifically detects CD18 in Rat samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.Specifications
CD18 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative | |
0 | |
ITGB2 | |
Rat beta2 Integrin | |
500 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Western Blot | |
WT.3 | |
Unconjugated | |
ITGB2 | |
2E6; AI528527; antigen CD18; B-2 integrin; CD18; CD18 leukocyte adhesion molecule; CD18 subunit; cell surface adhesion glycoprotein (LFA-1/CR3/P150,959 beta subunit precursor); cell surface adhesion glycoproteins LFA-1/CR3/p150,95 subunit beta; complement component 3 receptor 3 and 4 subunit; complement receptor C3 beta-subunit; complement receptor C3 subunit beta; integrin beta 2; integrin beta chain, beta 2; integrin beta-2; integrin subunit beta 2; integrin, beta 2; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; macrophage antigen 1 (mac-1) beta subunit); integrin, beta 2 (antigen CD18 subunit (p95), lymphocyte function-associated antigen 1, integrin B2); integrin, beta 2 (complement component 3 receptor 3 and 4 subunit); ITGB2; LAD; LCAMB; Leu-CAM receptor; leukocyte cell adhesion molecule CD18; leukocyte surface protein; leukocyte-associated antigens CD18/11A, CD18/11B, | |
Mouse | |
Protein G | |
RUO | |
309684 | |
-20°C | |
Liquid |