missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transferrin Monoclonal Antibody (A78-G/A7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA134862
Code : LE
Additional Details : Weight : 0.01000kg
Description
Suggested positive controls for this product are KG1 cells and human colorectal carcinoma tissues.
Transferrin Monoclonal antibody specifically detects Transferrin in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Transferrin | |
Monoclonal | |
0.2 mg/mL | |
10mM PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4 | |
P02787, P12346 | |
Tf | |
Neuraminidase-treated human red blood cells. | |
500 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgM |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
A78-G/A7 | |
Unconjugated | |
Tf | |
AI266983; beta-1 metal-binding globulin; Cd176; DKFZp781D0156; epididymis secretory sperm binding protein Li 71p; HEL-S-71p; HP; hpx; hypotransferrinemia with hemochromatosis; ICA; Inhibitor of carbonic anhydrase; lactoferrin; liver regeneration-related protein LRRG03; liver transferrin; LOC100146633; PICA; porcine inhibitor of carbonic anhydrase; porcine inhibitor of carbonic anhydrase precursor; PRO1400; PRO1557; PRO2086; Serotransferrin; siderophilin; TF; Tfn; TFQTL1; transferin; Transferrin; transferrin precursor; Trf | |
Mouse | |
Protein A | |
RUO | |
24825, 7018 | |
4° C, do not freeze | |
Liquid |