missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD17 Monoclonal Antibody (MEM-68), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110118
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD17 Monoclonal antibody specifically detects CD17 in Human samples. It is validated for Flow Cytometry.Specifications
CD17 | |
Monoclonal | |
1 mg/mL | |
TBS with 15mM sodium azide; pH 8 | |
Mouse | |
sequential chromatography | |
RUO | |
Human | |
Antibody | |
IgM |
Flow Cytometry | |
MEM-68 | |
Unconjugated | |
Lactosylceramide | |
Pre-B cell line NALM-6 | |
100 μg | |
Primary | |
4° C, do not freeze | |
Liquid |