Learn More
CD166 Monoclonal Antibody (105902), Alexa Fluor™ 488, Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA523565
Code : NEW
Additional Details : Weight : 0.00010kg
Description
This antibody shows approximately 50% cross-reactivity with recombinant mouse OCAM and no cross-reactivity with recombinant human (rh) BCAM, rhEpCAM, rhMCAM, or rhNCAM-L1.
CD166 Monoclonal antibody specifically detects CD166 in Human samples. It is validated for Flow Cytometry.Specifications
CD166 | |
Monoclonal | |
Alexa Fluor 488 | |
ALCAM | |
activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product | |
Mouse | |
Protein A/G | |
RUO | |
214 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
105902 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
Q13740 | |
ALCAM | |
Mouse myeloma cell line NS0-derived recombinant human ALCAM/CD166 Trp28-Ala526 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |