Learn More
CD163 Monoclonal Antibody (10D6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511458
Code : A0
Additional Details : Weight : 0.01000kg
Descripción
MA5-11458 targets CD163 in WB and IHC (P) applications and shows reactivity with Human samples. The MA5-11458 immunogen is recombinant protein encoding of domains 1-4 of the N-terminal region of human CD163.
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
CD163 | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
Q86VB7 | |
CD163 | |
Recombinant protein encoding of domains 1-4 of the N-terminal region of human CD163 | |
RUO | |
9332 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
10D6 | |
Unconjugated | |
CD163 | |
CD163; CD163 antigen; CD163 molecule; CD163v2; CD163v3; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; MM130; putative CD163 antigen; SCARI1; Scavenger receptor cysteine-rich type 1 protein M130; sCD163; Soluble CD163; Soluble sCD163 | |
Mouse | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |