missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD162 Monoclonal Antibody (TC2), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110117
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD162 Monoclonal antibody specifically detects CD162 in Human samples. It is validated for Flow Cytometry.Specifications
CD162 | |
Monoclonal | |
PE | |
SELPLG | |
CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
6404 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
TC2 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q14242 | |
SELPLG | |
Human thymocytes | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |