Learn More
KLRB1 Monoclonal Antibody (B199.2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181379
Code : LD
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD161 antibody, clone B199.2 recognizes the human Killer cell lectin-like receptor subfamily B member 1, also known as CD161, C-type lectin domain family 5 member B, HNKR-P1a, NKR-P1A or Natural killer cell surface protein P1A.
KLRB1 Monoclonal antibody specifically detects KLRB1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
KLRB1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q12918 | |
Klrb1 | |
Proprietary Immunogen. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
B199.2 | |
Unconjugated | |
Klrb1 | |
4930431A04Rik; Antigen 3.2.3; CD161; CD161 antigen-like family member A; CD161 antigen-like family member B; CD161a; CD161b; CLEC5B; C-type lectin domain family 5 member B; EMBL:AAQ11375.1}; Gm4696; HNKR-P1a; Immunoreceptor NKR-P1E; immunoreceptor NKR-P1G; inhibitory receptor NKR-P1B; killer cell lectin like receptor B1; killer cell lectin-like receptor subfamily A member 1 complex; killer cell lectin-like receptor subfamily B member 1; killer cell lectin-like receptor subfamily B member 1 complex; killer cell lectin-like receptor subfamily B member 1A; killer cell lectin-like receptor subfamily B member 1B; killer cell lectin-like receptor subfamily B member 1B allele A; Killer cell lectin-like receptor subfamily B member 1B allele B; killer cell lectin-like receptor subfamily B member 1B allele C; killer cell lectin-like receptor subfamily B member 1D; killer cell lectin-like receptor subfamily B member 1F; killer cell lectin-like receptor subfamily B member 1G; killer cell lectin-li | |
Mouse | |
Protein A | |
RUO | |
3820 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |