missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD160 Monoclonal Antibody (CNX46-3), Biotin, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517966
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17966 targets CD160/BY55 antigen in FACS and WB applications and shows reactivity with Mouse samples. The MA5-17966 immunogen is mouse CD160.
CD160 Monoclonal antibody specifically detects CD160 in Mouse samples. It is validated for Flow Cytometry, Western Blot.Specifications
CD160 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
O88875 | |
CD160 | |
Mouse CD160 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
CNX46-3 | |
Biotin | |
CD160 | |
AU045688; BY55; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 molecule; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell BY55; natural killer cell receptor; natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member; NK1; NK28; RGD1563598 | |
Rat | |
Protein G | |
RUO | |
54215 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |