missing translation for 'onlineSavingsMsg'
Learn More

CD16, Alexa Fluor 488, clone: 3G8, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA518127

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15347394

  • £286.16 / 100 tests


Estimated Shipment
08-05-2018
Add to basket

Specifications

Specifications

CD16
Monoclonal
Alexa Fluor 488
FCGR3A
Purified
FCGR3A, FCGR3B
Human neutrophils
Size-exclusion chromatography
DO NOT FREEZE!, Store at 4° C, store in dark
Primary
2214, 2215
Flow Cytometry
3G8
PBS with 0.2% BSA and 0.02% sodium azide
O75015, P08637
FCGR3A, FCGR3B, CD16A, CD16B, Fc gamma Receptor III, Fc gamma Receptor 3, RP11-5K23.1, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA
Murine
IgG1
100 Tests
Antibody
Monoclonal
Non-human Primate
Videos