Learn More
CD16 Monoclonal Antibody (ASH 1975), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17633
256.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-7633 detects CD16 from human samples and CD16-2 from mouse and rat. MA1-7633 has been successfully used in ELISA, IF, flow cytometry, immunoprecipitation, and immunohistochemistry (frozen sections) procedures. The MA1-7633 immunogen is a CD16 protein. Store at 4oC short term. For extended storage, store at -80oC.
CD16 Monoclonal antibody specifically detects CD16 in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
CD16 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
A0A0B4J1G0, O75015, P08508, P08637 | |
Fcgr3, FCGR3A, FCGR3B, Fcgr4 | |
CD16, Human white blood cell fraction | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
ASH 1975 | |
Unconjugated | |
FCGR3A | |
4833442P21Rik; CD16; CD16-2; CD16A; CD16a antigen; CD16b; CD32; CD32 receptor 2; CDw32; cytolytic trigger molecule G7; Fc fragment of IgG intermediate affinity IV receptor; Fc fragment of IgG low affinity IIIa receptor; Fc fragment of IgG receptor IIIa; Fc fragment of IgG receptor IIIb; Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor; Fc fragment of IgG, low affinity IIIa, receptor (CD16a); Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIb, receptor (CD16b); Fc gamma receptor III; Fc gamma receptor IIIa; Fc gamma receptor III-A; Fc gamma receptor IIIb; Fc gamma RIIIa; Fc gammaRIV; Fc receptor, IgG, low affinity III; Fc receptor, IgG, low affinity IV; Fc receptor-like 3; Fcg receptor III; FCG2; FCG3; Fc-gamma receptor III-2 (CD 16); Fc-gamma receptor IIIb (CD 16); | |
Mouse | |
Protein G | |
RUO | |
14131, 2214, 2215, 246256 | |
4°C short term, -80°C long term | |
Liquid |