Learn More
CD155 Monoclonal Antibody (D171), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513490
286.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-13490 targets CD155 in FACS, WB and ICC/IF applications and shows reactivity with Human, Mouse and Non-human primate samples. The MA5-13490 immunogen is heLa cells.
CD155 Monoclonal antibody specifically detects CD155 in Human, Mouse, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
CD155 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P15151 | |
PVR | |
HeLa cells | |
500 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
D171 | |
Biotin | |
PVR | |
3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4 | |
Mouse | |
Protein G | |
RUO | |
52118, 5817 | |
4° C | |
Liquid |