missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTLA-4 Monoclonal Antibody (WKH 203), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517587
323.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Rat samples. It is validated for Flow Cytometry.Specifications
CTLA-4 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
0 | |
CTLA4 | |
Purified rCTLA-4hlg fusion protein | |
200 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
WKH 203 | |
PE | |
CTLA4 | |
ALPS5; CD; CD152; CD152 antigen; CD152 isoform; CD152 protein; CD152 protein precursor; celiac disease 3; CELIAC3; Ctla4; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; cytotoxic T-lymphocyte associated protein 4; cytotoxic T-lymphocyte protein 4; cytotoxic T-lymphocyte protein 4 isoform CTLA4-TM; cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; EGK_04712; GRD4; GSE; ICOS; IDDM12; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; Ly-56; sCTLA4; soluble form; transmembrane form | |
Mouse | |
Protein G | |
RUO | |
63835 | |
4° C, do not freeze | |
Liquid |