missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD151 Monoclonal Antibody (IIG5a), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA190964
Code : LE
Additional Details : Weight : 0.01000kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD151 Monoclonal antibody specifically detects CD151 in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
CD151 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P48509 | |
CD151 | |
Proprietary immunogen - Contact ABR for details. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
IIG5a | |
Unconjugated | |
CD151 | |
CD 151; CD151; CD151 antigen; CD151 antigen (Raph blood group); CD151 molecule; CD151 molecule (Raph blood group); GP27; hemidesmosomal tetraspanin CD151; membrane glycoprotein SFA-1; MER2; Peta3; PETA-3; platelet endothelial tetraspan antigen-3; platelet surface glycoprotein gp27; platelet-endothelial cell tetraspan antigen 3; platelet-endothelial tetraspan antigen 3; RAPH; SFA1; SFA-1; Tetraspanin-24; TSPAN24; tspan-24 | |
Mouse | |
Protein A | |
RUO | |
977 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |