Learn More
CD150 Mouse anti-Human, Unlabeled, Clone: A12, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559591
Additional Details : Weight : 0.01000kg
Description
The A12 monoclonal antibody specifically binds to CD150, a single chain type I transmembrane glycoprotein of approximately 75-95 kDa. CD150 is encoded by the SLAMF1 gene and is also known as a signaling lymphocyte activation molecule (SLAM), or IPO-3. It is expressed on T cells, B cells, thymocytes, germinal center and dendritic cells, but not on monocytes. Expression of CD150 is upregulated on T, B and dendritic cells after activation via T-cell and B-cell costimulatory molecules. It has been reported that CD150 plays a role in signal transduction.
Specifications
CD150 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Activated Human Peripheral Blood Mononuclear Cells | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
A12 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |