Learn More
CD147 (soluble) Monoclonal Antibody (MEM-M6/1), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110104
258.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes an epitope in the N-terminal Ig domain (D1) of CD147 (Neurothelin), a 50-60 kDa type I transmembrane glycoprotein primarily expressed on all leukocytes, red blood cells, platelets and endothelial cells; it is not expressed by resting lymphocytes. This is a high-affinity antibody capable of binding to unstimulated peripheral blood T cells.
CD147 (soluble) Monoclonal antibody specifically detects CD147 (soluble) in Human samples. It is validated for Flow Cytometry.Specifications
CD147 (soluble) | |
Monoclonal | |
APC | |
BSG | |
5A11; 5A11 antigen; 5A11/Basigin; 5A11/Basigin-2; 5F7; AI115436; AI325119; Basic immunoglobulin superfamily; basigin; basigin (Ok blood group); Basigin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); basignin; Basignin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); blood-brain barrier HT7 antigen; BSG; CD147; CD147 antigen; CE9; Collagenase stimulatory factor; EMMPRIN; Extracellular matrix metalloproteinase inducer; glycoprotein CE9; gp 42; gp42/basigin/OX-47/HT7; HT7; HT-7; HT7 antigen; immunoglobulin gene superfamily member; leukocyte activation antigen M6; Membrane glycoprotein gp42; neurothelin; OK; OK blood group antigen; OX-47 antigen; Ox47R; RPE7; TCSF; tumor cell-derived collagenase stimulatory factor; Unknown (protein for MGC:128392); unnamed protein product; UNQ6505/PRO21383 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
682 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-M6/1 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P35613 | |
BSG | |
Protein A-CR purified soluble recombinant form of CD147, CD147Rg, which consists of the cDNA coding for the hinge region, CH2-and CH3 domain of human IgG1 (CD147Rg is secreted by transfectants as a dimer). | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |