Learn More
CD146 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528893
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HUVEC, rat liver extract. Predicted reactivity: Mouse (81%), Rat (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD146 Polyclonal antibody specifically detects CD146 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD146 | |
Polyclonal | |
Unconjugated | |
MCAM | |
1-gicerin; AV025631; CD146; CD149; Cell surface glycoprotein MUC18; cell surface glycoprotein P1H12; Gicerin; HEMCAM; l-gicerin; l-gicerin precursor; l-gicerin protein; LOW QUALITY PROTEIN: cell surface glycoprotein MUC18; MCAM; melanoma adhesion molecule; melanoma cell adhesion molecule; Melanoma-associated antigen A32; melanoma-associated antigen MUC18; Muc18; mucin-18; s-endo; S-endo 1 endothelial-associated antigen; s-gicerin; s-gicerin precursor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4162, 78967, 84004 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P43121, Q8R2Y2, Q9EPF2 | |
MCAM | |
Recombinant fragment corresponding to a region within amino acids 421 and 646 of Human CD146 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |