missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFRA Monoclonal Antibody (16A1), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110097
Code : LD
Additional Details : Weight : 0.01000kg
Description
PDGFRA Monoclonal antibody specifically detects PDGFRA in Human samples. It is validated for Flow Cytometry.Specifications
PDGFRA | |
Monoclonal | |
APC | |
PDGFRA | |
AI115593; alpha platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor precursor (PDGF-R-alpha); alpha-type platelet-derived growth factor receptor; APDGFR; CD140 antigen-like family member A; CD140A; CD140a antigen; GAS9; MGC74795; OTTHUMP00000218656; PDGF A-chain; PDGF alpha chain; PDGF Receptor alpha; PDGF subunit A; PDGF-1; PDGFACE; PDGFR2; Pdgfr-2; PDGFRA; pdgfr-a; PDGFRA/BCR fusion; PDGF-R-alpha; PDGFR-alpha; platelet derived growth factor receptor alpha; platelet derived growth factor receptor, alpha polypeptide; platelet-derived growth factor alpha receptor; Platelet-derived growth factor receptor 2; platelet-derived growth factor receptor alpha; platelet-derived growth factor receptor, alpha polypeptide; rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein; RHEPDGFRA | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
5156 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
16A1 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P16234 | |
PDGFRA | |
CD140a-transfected NIH 3T3 cells | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |