Learn More
CD14 Monoclonal Antibody (TuK4), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA182234
286.68 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti human CD14 antibody, clone TÜK4 recognizes the human CD14 cell surface antigen.
CD14 Monoclonal antibody specifically detects CD14 in Bovine, Camelid, Canine, Cynomolgus Monkey, Feline, Goat, Human, Mink, Ovine, Porcine, Rabbit samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
P08571, Q28680, Q95122 | |
Cd14 | |
Proprietary Immunogen. | |
100 Tests | |
Primary | |
Bovine, Camelid, Camelid, Cynomolgus Monkey, Cynomolgus Monkey, Goat, Goat, Mink, Mink, Porcine, Porcine | |
Antibody | |
IgG2a |
Flow Cytometry | |
TuK4 | |
PE | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
Protein A | |
RUO | |
100008983, 100037938, 100861226, 101096132, 102129342, 102177056, 281048, 443216, 607076, 929 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |