missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513306
Code : LE
Additional Details : Weight : 0.01000kg
Description
CD14 Polyclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD14 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P08571 | |
Cd14 | |
KLH conjugated synthetic peptide between 292-322 amino acids from the C-terminal region of human CD14 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
929 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |