Learn More
CD14 Monoclonal Antibody (MEM-18), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119561
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD14 is a 55kDa GPI-anchored glycoprotein, constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils, where serves as a multifunctional lipopolysaccharide receptor; it is also released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 is able to discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. CD14 affects allergic, inflammatory and infectious processes.
CD14 Monoclonal antibody specifically detects CD14 in Human, Non-human Primate samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Monoclonal | |
FITC | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
929 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-18 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P08571 | |
Cd14 | |
Human urinary proteins from the urine of a patient suffering from proteinuria. | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |