Learn More
CD14 Monoclonal Antibody (18D11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA140324
Code : LD
Additional Details : Weight : 0.10000kg
Description
MA1-40324 detects CD14 from human samples. MA1-40324 has been successfully used in functional assays and flow cytometry applications. The MA1-40324 immunogen is CD14. Endotoxin <24 EU/mg.
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry, Functional Assay.Specifications
CD14 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and no preservative | |
P08571 | |
Cd14 | |
CD14 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay | |
18D11 | |
Unconjugated | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
Protein G | |
RUO | |
929 | |
4° C | |
Liquid |