Learn More
CD138 Monoclonal Antibody (B-A38), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181808
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti human CD138 antibody, clone B-A38 recognizes human CD138, also known as Syndecan-1 (SDC-1).
CD138 Monoclonal antibody specifically detects CD138 in Human samples. It is validated for Flow Cytometry.Specifications
CD138 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
P18827 | |
Sdc1 | |
U266 cell line. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
B-A38 | |
PE | |
Sdc1 | |
AA408134; AA409076; CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; HSPG; sCD138; SDC; Sdc1; soluble CD138; Sstn; syn-1; Synd; SYND1; Synd-1; SYNDECA; syndecan; syndecan 1; syndecan proteoglycan 1; Syndecan1; syndecan-1; synstatin | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
6382 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |