missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD137 Monoclonal Antibody (BBK-2), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513736
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-13736 targets CD137 in FACS and IF applications and shows reactivity with Human samples. The MA5-13736 immunogen is ectodomain of human 4-1BB recombinant protein.
CD137 Monoclonal antibody specifically detects CD137 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
CD137 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Q07011 | |
TNFRSF9 | |
Ectodomain of human 4-1BB recombinant protein | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry | |
BBK-2 | |
Biotin | |
TNFRSF9 | |
4 1 BB; 41BB; 4-1BB; 4-1BB ligand receptor; A930040I11Rik; AA408498; AI325004; Cd137; CD137 antigen; CDw137; EGK_00183; homolog of mouse 4-1BB; ILA; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; Ly63; receptor protein 4-1BB; s4 1 BB; s4 1BB; s41BB; sCD137; secreted CD137 antigen; soluble 4 1 BB; soluble 4 1BB; soluble 41BB; soluble CD137; T cell antigen ILA; T-cell antigen 4-1BB; T-cell antigen 4-1BB homolog; T-cell antigen ILA; TNF receptor superfamily member 9; TNFRSF9; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9 | |
Mouse | |
Protein G | |
RUO | |
3604 | |
4° C | |
Liquid |