missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40L Monoclonal Antibody (OX89), Biotin, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517911
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17911 targets CD134L/OX40L in FACS and IHC applications and shows reactivity with Mouse samples. The MA5-17911 immunogen is cD4.CD134L chimeric protein.
OX40L Monoclonal antibody specifically detects OX40L in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
OX40L | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P43488 | |
Tnfsf4 | |
CD4.CD134L chimeric protein | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry | |
OX89 | |
Biotin | |
Tnfsf4 | |
Ath1; Ath-1; atherosclerosis 1; CD134 ligand; CD134L; CD252; glycoprotein Gp34; GP34; OX40 antigen ligand; OX40 ligand; OX40L; OX-40L; OX4OL; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 ligand; TNFSF4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor (ligand) superfamily, member 4; tumor necrosis factor ligand 2B; tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; TXGP1; Txgp1l | |
Rat | |
Protein G | |
RUO | |
22164 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |