missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535326
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CD11c Polyclonal antibody specifically detects CD11c in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD11c | |
Polyclonal | |
Unconjugated | |
Itgax | |
AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3687 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P20702 | |
Itgax | |
KLH conjugated synthetic peptide between 1119-1145 amino acids from the C-terminal region of human ITGAX | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |