Learn More
CD11c Monoclonal Antibody (N418), Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen MA180129
144.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Hamster anti Mouse CD11c antibody, clone N418 recognizes the murine homolog of human CD11c, also known as Integrin Alpha X, a 150/90 kDa member of the beta 2 integrin family.
CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
CD11c | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9QXH4 | |
Itgax | |
Murine spleen dendritic cells. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
N418 | |
Unconjugated | |
Itgax | |
AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6 | |
Armenian Hamster | |
Protein G | |
RUO | |
16411 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |