Learn More
anti-CD11b, Polyclonal, Abcam™
Brand: Abcam AB75476-100UL
Code : Z1
Additional Details : Weight : 0.25000kg
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Cow, Human. Predicted to work with Sheep
Immunogen: A synthetic peptide from an internal region (within residues 250-350) of mouse CD11b. [Swiss-Prot P05555]
Applications:
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting
Specifications
CD11b | |
1mg/mL | |
Rabbit | |
Affinity Purified | |
Signal Transduction, Stem Cell Biology, Tags And Cell Markers | |
Bovine, Human, Mouse, Rat, Sheep |
Polyclonal | |
Unconjugated | |
A synthetic peptide from an internal region (within residues 250-350) of mouse CD11b. [Swiss-Prot P05555] | |
100 μg | |
Primary | |
IgG |