Learn More
CD11b/c Monoclonal Antibody (OX-42), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181606
Code : LD
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat CD11b, clone OX-42, recognizes rat CD11b, also known as integrin alpha-M, the receptor for the iC3b component of complement. CD11b is a 1151 amino acid single pass type 1 transmembrane glycoprotein posessing a single vWFA domain and multiple FG-GAP repeats. Mouse anti Rat CD11b, clone OX-42, recognizes rat CD11b, also known as integrin alpha-M, the receptor for the iC3b component of complement. CD11b is a 1151 amino acid single pass type 1 transmembrane glycoprotein posessing a single vWFA domain and multiple FG-GAP repeats.
CD11b/c Monoclonal antibody specifically detects CD11b/c in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
CD11b/c | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
0 | |
Itgam, Itgax | |
Rat peritoneal macrophages. | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
OX-42 | |
Unconjugated | |
Itgam | |
AI449405; antigen CD11b (p170); CD11 antigen-like family member B; CD11 antigen-like family member C; CD11b; CD11B (p170); CD11b/CD18; CD11bc; CD11C; CD11C (p150) alpha polypeptide; cell surface glycoprotein MAC-1 alpha subunit; cell surface glycoprotein MAC-1 subunit alpha; complement component 3 receptor 3 subunit; complement component 3 receptor 4 subunit; complement component receptor 3 alpha-a; complement receptor 4; complement receptor type 3; CR3; CR-3 alpha chain; CR3A; Cr4; F730045J24Rik; integrin alpha M; integrin alpha X; integrin alpha-M; Integrin alpha-X; integrin aX; integrin subunit alpha M; integrin subunit alpha X; integrin, alpha M; integrin, alpha M (complement component 3 receptor 3 subunit); integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); ITGAM; ITGAX; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor MO1; leuk | |
Mouse | |
Protein A | |
RUO | |
25021, 499271 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |