Learn More
CD11a Monoclonal Antibody (121/7), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA11A5
Code : PN
Additional Details : Weight : 0.80000kg
Description
MA11A5 targets CD11a in FACS, IA, and Neu applications and shows reactivity with mouse samples. This antibody does not react with human Jurkat cells in FACS applications. The MA11A5 immunogen is mouse 180kD alpha chain (CD11alpha) of the integrin Lymphocyte Function-Associated Antigen-1. MA11A5 detects CD11a which has a predicted molecular weight of approximately 126 kDa. This antibody is produced by injecting Rat IgG secreting hybridoma cells into the peritoneum of mice. The resulting ascites is collected from the mice and the antibody is purified. This product is a Low Endotoxin formulation. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
CD11a Monoclonal antibody specifically detects CD11a in Mouse samples. It is validated for Flow Cytometry, Inhibition Assays, Neutralization.Specifications
CD11a | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P24063 | |
ITGAL | |
Mouse 180kD alpha chain (CD11alpha) of the integrin Lymphocyte Function-Associated Antigen-1 | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry, Inhibition Assays, Neutralization | |
121/7 | |
Unconjugated | |
ITGAL | |
(p180); alpha L integrin; antigen CD11A (p180); antigen CD11A (p180), lymphocyte function-associated antigen 1, alpha polypeptide; antigen CD11A p180; CD11 antigen-like family member A; CD11a; integrin alpha L; integrin alphaL; integrin alpha-L; integrin alpha-L precursor; integrin gene promoter; integrin subunit alpha L; integrin surface receptor; integrin, alpha L; integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide); ITGAL; Leukocyte adhesion glycoprotein LFA-1 alpha chain; leukocyte function-associated molecule 1 alpha chain; LFA-1; LFA-1 alpha; LFA1A; LFA-1A; Ly-15; Ly-21; lymphocyte antigen 15; lymphocyte function associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen 1; lymphocyte function-associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen-1 | |
Rat | |
Protein G | |
RUO | |
16408 | |
-20°C | |
Liquid |