missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD109 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA524136
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD109 Polyclonal antibody specifically detects CD109 in Human, Mouse samples. It is validated for Western Blot.Specifications
CD109 | |
Polyclonal | |
Unconjugated | |
CD109 | |
150 kDa TGF-beta-1-binding protein; 9930012E15Rik; activated T-cell marker CD109; AI480638; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; Cd109; CD109 antigen; CD109 molecule; CPAMD7; GARP; Gov platelet alloantigens; GPI-anchored alpha 2 macroglobulin-related protein; GPI-anchored alpha-2 macroglobulin-related protein; p180; platelet-specific Gov antigen; r150; RP11-525G3.1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
135228, 235505 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q6YHK3, Q8R422 | |
CD109 | |
KLH conjugated synthetic peptide between 82-109 amino acids from the N-terminal region of human CD109 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |