missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCAM-1 Monoclonal Antibody (STA), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110068
218.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
VCAM-1 Monoclonal antibody specifically detects VCAM-1 in Human samples. It is validated for Flow Cytometry.Specifications
VCAM-1 | |
Monoclonal | |
PE | |
VCAM1 | |
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
7412 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
STA | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P19320 | |
VCAM1 | |
Human DS6 T cell line | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |